missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERLIN1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA519152
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
ERLIN1 Polyclonal antibody specifically detects ERLIN1 in Human samples. It is validated for Western Blot.Spezifikation
ERLIN1 | |
Polyclonal | |
Unconjugated | |
Erlin1 | |
2810439N09Rik; Band_7 23-211 Keo4 (Interim) similar to C.elegans protein C42C1.9; C10orf69; C80197; endoplasmic reticulum lipid raft-associated protein 1; ER lipid raft associated 1; Erlin1; erlin-1; KE04; Keo4; OTTHUMP00000059425; OTTHUMP00000197867; Protein KE04; Protein KE04 homolog; RGD1307058; SPFH domain family, member 1; SPFH domain-containing protein 1; Spfh1; SPG62; stomatin-prohibitin-flotillin-HflC/K domain-containing protein 1 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
10613 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O75477 | |
Erlin1 | |
Synthetic peptide sequence (EALEPSGENVIQNKE) corresponding to the C-terminus amino acids of ERLIN1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |