Learn More
Cytokeratin 36 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531829
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa. Predicted reactivity: Mouse (80%), Rat (82%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Cytokeratin 36 Polyclonal antibody specifically detects Cytokeratin 36 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Cytokeratin 36 | |
Polyclonal | |
Unconjugated | |
KRT36 | |
HA6; hair keratin; Hair keratin, type I Ha6; hHa6; HKA6; Hra-1; K36; Ka31; keratin 36; keratin 36, type I; keratin complex 1, acidic, gene 5; keratin, hair, acidic, 6; keratin, type I cuticular Ha6; Keratin-36; keratin-5; KRT hHa6; Krt1-22; Krt1-5; Krt36; KRTHA6; MHRa-1; type I hair keratin KA31; Type I keratin 48 kDa; type I keratin 48 kDa (MHRa-1) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8689 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
O76013 | |
KRT36 | |
Recombinant fragment corresponding to a region within amino acids 258 and 467 of Human Keratin 36 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |