Learn More
Ki-67 Monoclonal Antibody (4A1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA515525
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA5-15525 targets KI67 in indirect ELISA, IHC applications and shows reactivity with Human samples. The MA5-15525 immunogen is synthetic peptide corresponding to aa (CEDLAGFKELFQTPG) of human KI67, conjugated to KLH. MA5-15525 detects KI67 which has a predicted molecular weight of approximately 358kDa.
Ki-67 Monoclonal antibody specifically detects Ki-67 in Human samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry.Spezifikation
Ki-67 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P46013 | |
Mki67 | |
Synthetic peptide corresponding to aa (CEDLAGFKELFQTPG) of human KI67, conjugated to KLH. | |
RUO | |
4288 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry | |
4A1 | |
Unconjugated | |
Mki67 | |
antigen identified by monoclonal antibody Ki 67; antigen identified by monoclonal antibody Ki-67; Antigen identified by monoclonal antibody Ki-67 homolog; Antigen KI-67; Antigen KI-67 homolog; antigen KI-67; proliferation marker protein Ki-67; antigen KI-67-like; cb31; D630048A14Rik; I79_022666; Ki67; Ki-67; KIA; LOW QUALITY PROTEIN: proliferation marker protein Ki-67; marker of proliferation Ki-67; MIB-; MIB-1; Mki67; PPP1R105; Proliferation marker protein Ki-67; proliferation-related Ki-67 antigen; protein phosphatase 1, regulatory subunit 105; RP11-380J17.2; sb:cb31; si:ch211-250b22.7; unnamed protein product; wu:fa11g09; wu:fb57a07; wu:fi14e05 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |