missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIF7 Monoclonal Antibody (3F8), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA518325
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
Do not aliquot.
This gene encodes a cilia-associated protein belonging to the kinesin family. This protein plays a role in the sonic hedgehog (SHH) signaling pathway through the regulation of GLI transcription factors. It functions as a negative regulator of the SHH pathway by preventing inappropriate activation of GLI2 in the absence of ligand, and as a positive regulator by preventing the processing of GLI3 into its repressor form. Mutations in this gene have been associated with various ciliopathies. [provided by RefSeq, Oct 2011].Spezifikation
KIF7 | |
Monoclonal | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q2M1P5, B7ZNG0 | |
Mouse | |
Affinity chromatography | |
RUO | |
16576, 293047, 374654 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
3F8 | |
Unconjugated | |
KIF7 | |
KIF7 | |
A synthetic peptide derived from human Kif 7 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |