Learn More
Kinesin 5A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA1642
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-642 detects kinesin 5A from mouse, human, and bovine samples. This antibody is specific for kinesin 5A and does not detect other kinesin isotypes. PA1-642 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ∽110 kDa protein representing kinesin 5A from mouse neuronal cell lysate and human retinal extracts. The PA1-642 immunizing peptide corresponds to amino acid residues 1007-1027 from mouse kinesin 5A. This sequence is completely conserved in human. This peptide (Cat. # PEP-197) is available for use in neutralization and control experiments.
Kinesin 5A Polyclonal antibody specifically detects Kinesin 5A in Bovine, Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
Kinesin 5A | |
Polyclonal | |
Unconjugated | |
KIF5A | |
D10Bwg0738e; D12S1889; Khc; Kiaa4086; Kif5; Kif5a; kif5a {ECO:0000312; KIF5A variant protein; kinesin family member 5A; kinesin heavy chain isoform 5A; kinesin heavy chain isoform 5A; kinesin heavy chain; kinesin heavy chain neuron-specific 1; kinesin, heavy chain, neuron-specific; Kns; mKIAA4086; MY050; Neuronal kinesin heavy chain; NKHC; Nkhc1; RGD:1303035}; SPG10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16572, 3798, 511273 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P33175, Q12840 | |
KIF5A | |
Synthetic Peptide: C(1007) G Y E A E D Q A K L F P L H Q E T A A S(1027) | |
100 μg | |
Primary | |
Bovine, Human, Mouse | |
Antibody | |
IgG |