missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kinesin 5B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA119392
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody detects total levels of endogenous kinesin protein.
Kinesin 5B Polyclonal antibody specifically detects Kinesin 5B in Human, Mouse, Porcine samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
Kinesin 5B | |
Polyclonal | |
Unconjugated | |
KIF5B | |
AL022807; Conventional kinesin heavy chain; EMBL:ABC25059.1}; epididymis secretory protein Li 61; HEL-S-61; Khc; Khcs; KIF5A; KIF5B; kif5b {ECO:0000312; kinesin 1 (110-120kD); kinesin family member 5A; kinesin family member 5B; kinesin heavy chain; kinesin heavy chain member 5B, ubiquitous; kinesin-1 heavy chain; KINH; KNS; Kns1; ubiquitous kinesin heavy chain; UKHC; U-KHC; Unknown (protein for IMAGE:7944745) | |
Rabbit | |
Affinity chromatography | |
RUO | |
16573, 3799, 595132 | |
4° C, do not freeze | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P33176, Q61768 | |
KIF5B | |
Stalk domain of human kinesin, expressed in (aa 331-906) E. coli. | |
100 μg | |
Primary | |
Human, Mouse, Porcine | |
Antibody | |
IgG |