missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIR3DL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA526178
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
KIR3DL3 is transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. This protein are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13.4 within the 1 Mb leukocyte receptor complex (LRC).Spezifikation
KIR3DL3 | |
Polyclonal | |
Unconjugated | |
KIR3DL3 | |
CD158 antigen-like family member Z; CD158Z; killer cell Ig-like receptor KIR3DL7; killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 3; killer cell immunoglobulin-like receptor 3DL3; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 2; killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 3; killer cell inhibitory receptor 1; killer-cell immunoglobulin-like receptor 3DL3; KIR2DS2; KIR3DL3; KIR3DL7; KIR44; KIRC1; natural killer cell receptor | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
115653 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 0.09% sodium azide | |
Q8N743 | |
KIR3DL3 | |
KLH conjugated synthetic peptide between 132-158 amino acids from the central region of human KIR3DL3 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |