missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kir4.1 (KCNJ10) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA537137
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
This antibody detects endogenous protein at a molecular weight of 42 kDa. Purity is >95% by SDS-PAGE.
Kir4.1 (KCNJ10) Polyclonal antibody specifically detects Kir4.1 (KCNJ10) in Human, Mouse, Rat samples. It is validated for Western Blot.Spezifikation
Kir4.1 (KCNJ10) | |
Polyclonal | |
Unconjugated | |
KCNJ10 | |
ATP-dependent inwardly rectifying potassium channel Kir4.1; ATP-sensitive inward rectifier potassium channel 10; ATP-sensitive inward rectifier potassium channel KAB-2; BIR10; BIRK1; BIRK-1; BIRK10; BIRK-10; brain-specific inwardly rectifying K(+) channel 1; glial ATP-dependent inwardly rectifying potassium channel KIR4.1; inward rectifier K(+) channel Kir1.2; Inward rectifier K(+) channel Kir4.1; inward rectifier K+ channel KIR1.2; Kab-2; Kcnj10; KCNJ13 PEN; KCNJ13-PEN; Kir1.2; Kir4.1; potassium channel, inwardly rectifying subfamily J member 10; potassium channel, inwardly rectifying subfamily J, member 10; potassium inwardly-rectifying channel, subfamily J, member 10; potassium voltage-gated channel subfamily J member 10; SESAME | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16513, 29718, 3766 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P49655, P78508, Q9JM63 | |
KCNJ10 | |
A synthetic peptide corresponding to residues in human KIR41 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |