Learn More
KLHDC5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531632
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HepG2. Predicted reactivity: Mouse (98%), Rat (97%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Substrate-specific adapter of a BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex required for mitotic progression and cytokinesis. The BCR(KLHL42) E3 ubiquitin ligase complex mediates the ubiquitination and subsequent degradation of KATNA1. Involved in microtubule dynamics throughout mitosis. [UniProt]Spezifikation
KLHDC5 | |
Polyclonal | |
Unconjugated | |
KLHL42 | |
A330079N23Rik; AI450355; C230080I20Rik; Ctb9; cullin-3-binding protein 9; kelch domain containing 5; kelch domain-containing protein 5; kelch like family member 42; kelch-like 42; kelch-like family member 42; kelch-like family, member 42; kelch-like protein 42; KIAA1340; Klhdc5; KLHL42; mKIAA1340 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
232539, 57542 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8BFQ9, Q9P2K6 | |
KLHL42 | |
Recombinant fragment corresponding to a region within amino acids 170 and 488 of Human KLHDC5 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |