missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kallikrein 2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518674
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
Glandular kallikreins cleave Met-Lys and Arg-Ser bonds in kininogen to release Lys-bradykinin.Spezifikation
Kallikrein 2 | |
Polyclonal | |
Unconjugated | |
KLK2 | |
glandular kallikrein 2; glandular kallikrein-1; hGK-1; hK2; kallikrein 2, prostatic; kallikrein related peptidase 2; kallikrein-2; kallikrein-related peptidase 2; KLK2; KLK2A2; MGC12201; Tissue kallikrein 2; Tissue kallikrein-2 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
3817 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P20151 | |
KLK2 | |
Synthetic peptide sequence (KHQSLRPDEDSSHD) corresponding to the internal amino acids of KLK2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |