missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLRF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA543007
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: QKGSCSNATQ YEDTGDLKVN NGTRRNISNK DLCASRSADQ TVLCQSEWLK Sequence homology: Human: 100%
KLRF1 Polyclonal antibody specifically detects KLRF1 in Human samples. It is validated for Western Blot.Spezifikation
KLRF1 | |
Polyclonal | |
Unconjugated | |
KLRF1 | |
Activating coreceptor NKp80; CLEC5C; CLEC5CMGC119908; C-type lectin domain family 5 member C; killer cell lectin like receptor F1; killer cell lectin-like receptor F1; killer cell lectin-like receptor subfamily F member 1; killer cell lectin-like receptor subfamily F, member 1; KLRF1; Lectin-like receptor F1; MGC119907; MGC119909; ML; natural killer cell receptor; NKp80 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51348 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NZS2 | |
KLRF1 | |
Synthetic peptide directed towards the middle region of human KLRF1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |