missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAMP2 Monoclonal Antibody (M3/84), Invitrogen™
Rat Monoclonal Antibody
Marke: Invitrogen MA517861
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
LAMP2 Monoclonal antibody specifically detects LAMP2 in Mouse samples. It is validated for Cytotoxicity Assay, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Spezifikation
LAMP2 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P17047 | |
LAMP2 | |
Immunoadsorbent purified mouse macrophage glycoprotein fraction. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
Cytotoxicity Assay, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
M3/84 | |
Unconjugated | |
LAMP2 | |
CD107 antigen-like family member B; CD107b; EGK_20887; Igp96; Lamp II; LAMP2; LAMP-2; LAMP-2 Isoform 2A; LAMP-2 Variant A; Lamp-2a; Lamp-2b; Lamp-2c; LAMPB; LGP110; LGP-110; LGP-96; LGP-B; lysosomal associated membrane protein 2; lysosomal membrane glycoprotein 2; lysosomal membrane glycoprotein type B; lysosomal-associated membrane protein 2; lysosome-associated membrane glycoprotein 2; lysosome-associated membrane protein 2; Mac3; RP23-193O17.2 | |
Rat | |
Protein G | |
RUO | |
16784 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |