Learn More
LAMP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA1654A
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-654A detects Lamp1 from human samples. PA1-654A has been successfully used in immunofluorescence and Western blotting procedures. In immunofluorescence procedures, this antibody stains Lamp1 in HeLa cells resulting in perinuclear staining. The PA1-654A immunogen is a synthetic peptide corresponding to residues C K(407) R S H A G Y Q T I(416) of human LAMP1. This region is completely conserved between human, mouse and rat.
LAMP1 Polyclonal antibody specifically detects LAMP1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
LAMP1 | |
Polyclonal | |
Unconjugated | |
LAMP1 | |
120 kDa lysosomal membrane glycoprotein; AI196048; CD107 antigen-like family member A; CD107a; I79_011073; Lamp I; LAMP1; LAMP-1; LAMPA; LGP120; LGP-120; LGPA; LGP-A; lysosomal associated membrane protein 1; Lysosomal associated membrane protein 1 (120 kDa); lysosomal membrane glycoprotein 1; lysosomal membrane glycoprotein A; lysosomal-associated membrane protein 1; lysosome-associated membrane glycoprotein 1; LYSOSOME-ASSOCIATED MEMBRANE GLYCOPROTEIN 1 PRECURSOR (LAMP-1) (LGP-A) (LGP-120) (CD107A) (P2B); lysosome-associated membrane protein 1; P2B | |
Rabbit | |
Affinity chromatography | |
RUO | |
3916 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.4 | |
P11279 | |
LAMP1 | |
Synthetic peptide corresponding to residues C K(407) R S H A G Y Q T I(416) of human LAMP1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |