missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LCK Monoclonal Antibody (LCK-01), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA119197
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody will not cross-react with mouse.
LCK Monoclonal antibody specifically detects LCK in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot.Spezifikation
LCK | |
Monoclonal | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
P06239 | |
LCK | |
Peptide corresponding to amino acids 22-36 in the sequence of human Lck. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
LCK-01 | |
Unconjugated | |
LCK | |
EC 2.7.10.2; Hck-3; IMD22; kinase Lck; Lck; LCK proto-oncogene, Src family tyrosine kinase; Lck1; Lcktkr; leukocyte C-terminal Src kinase; LSK; Lskt; Lsk-t; Lymphocyte cell-specific protein-tyrosine kinase; lymphocyte protein tyrosine kinase; lymphocyte-specific protein tyrosine kinase; OTTHUMP00000008640; OTTHUMP00000008740; OTTHUMP00000008741; p56(LSTRA) protein-tyrosine kinase; p56<lck>; p56lck; p56-LCK; pp58lck; Protein YT16; Proto-oncogene Lck; Proto-oncogene tyrosine-protein kinase LCK; RP4-675E8.4; t cell-specific protein-tyrosine kinase; T-lymphocyte specific protein tyrosine kinase p56lck; tyr; tyrosine-protein kinase Lck; YT16 | |
Mouse | |
Protein A | |
RUO | |
3932 | |
4° C, do not freeze | |
Liquid |