missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGMN Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA547244
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
In direct ELISAs, approximately 50% cross-reactivity with recombinant human Legumain is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
LGMN Polyclonal antibody specifically detects LGMN in Mouse samples. It is validated for Western Blot.Spezifikation
LGMN | |
Polyclonal | |
Unconjugated | |
LGMN | |
AEP; AI746452; Asparaginyl endopeptidase; AU022324; cb173; cb460; cysteine protease 1; legumain; Lgmn; LGMN1; preprolegumain; protease, cysteine 1; protease, cysteine, 1; protease, cysteine, 1 (legumain); PRSC1; PRSC1EC 3.4.22.34; sb:cb173; wu:fa98g05; wu:fj92g08; zgc:76953 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
19141 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O89017 | |
LGMN | |
Mouse myeloma cell line NS0-derived recombinant mouse Legumain Val18-Tyr435 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |