missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lidocaine Monoclonal Antibody (LC11-222.4), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA110752
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-10752 detects Lidocaine. MA1-10752 has been successfully used in ELISA applications. The MA1-10752 immunogen is Lidocaine, conjugated to KLH.
Lidocaine is a synthetic aminoethylamide with local anesthetic and antiarrhythmic properties. Lidocaine stabilizes the neuronal membrane by binding to and inhibiting voltage-gated sodium channels, thereby inhibiting the ionic fluxes required for the initiation and conduction of impulses and effecting local anesthesia. [National Center for Biotechnology Information. PubChem Compound Database; CID=3676].Spezifikation
Lidocaine | |
Monoclonal | |
1.16 mg/mL | |
PBS with no preservative; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG1 |
ELISA | |
LC11-222.4 | |
Unconjugated | |
Anestacon; C14H22N2O; Lidoderm; Lignocaine; Xylocaine | |
Lidocaine, conjugated to KLH. | |
500 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |