Learn More
Listeria Polyclonal Antibody, FITC, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA173128
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The PA1-73128 antibody reacts with Listeria species. PA1-73128 has been successfully used in immunofluorescence applications. Acetone fixation is recommended prior to staining. The PA1-73128 antibody was raised against Listeria monocytogenes; ATCC strain 43251. Product can be stored at 4oC for up to 6 months. For long term storage, please aliquot and store at -20oC, avoiding freeze/thaw cycles. Store in the dark.
Listeria Polyclonal antibody specifically detects Listeria in Bacteria samples. It is validated for Immunocytochemistry.Spezifikation
Listeria | |
Polyclonal | |
FITC | |
L. monocytogenes | |
Listeria monocytogenes; ATCC strain 43251. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |
Immunocytochemistry | |
4-5 mg/mL | |
PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 | |
Rabbit | |
purified | |
RUO | |
Bacteria | |
Antibody | |
IgG |