missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOX Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA116955
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is 92% homologous to chicken, zebrafish, and xenopus.
LOX Polyclonal antibody specifically detects LOX in Bovine, Human, Mouse, Porcine, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
LOX | |
Polyclonal | |
Unconjugated | |
LOX | |
AI893619; cross-linking enzyme; H-rev142; LOX; Lysyl oxidase; Lysyl oxidase (an H-rev gene with its expression down-regulated in HRAS-transformed rat 208F fibroblasts); MGC105112; protein-lysine 6-oxidase; Protein-lysine 6-oxidase, long form; Protein-lysine 6-oxidase, short form; Ras excision protein; ras recision; rrg; Rrg1; TSC-160 | |
Rabbit | |
Affinity chromatography | |
RUO | |
100525278, 16948, 24914, 280841, 4015 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P16636, P28300, P28301, P33072, P45845 | |
LOX | |
Cocktail of two synthetic peptides between amino acids 300-350 and amino acids 200-300 of the human LOX protein. | |
50 μL | |
Primary | |
Bovine, Human, Mouse, Porcine, Rat | |
Antibody | |
IgG |