missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPGAT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA545334
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: DQQLLLLKKH LENNYRSRDR KWIVLFPEGG FLRKRRETSQ AFAKKNNLPF
LPGAT1 Polyclonal antibody specifically detects LPGAT1 in Human samples. It is validated for Western Blot.Spezifikation
LPGAT1 | |
Polyclonal | |
Unconjugated | |
LPGAT1 | |
Acyl-CoA:lysophosphatidylglycerol acyltransferase 1; AI649174; AW112037; BC013667; FAM34A; FAM34A1; family with sequence similarity 34, member A; KIAA0205; Lpgat1; lysophosphatidylglycerol acyltransferase 1; NET8 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9926 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q92604 | |
LPGAT1 | |
Synthetic peptide directed towards the middle region of human LPGAT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |