missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRP4 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518999
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
LRP4 Polyclonal antibody specifically detects LRP4 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
LRP4 | |
Polyclonal | |
Unconjugated | |
LRP4 | |
6430526J12Rik; CLSS; CMS17; Corin; D230026E03; Kiaa0816; LDL receptor related protein 4; LDLR; LDLR dan; low density lipoprotein receptor-related protein 4; low-density lipoprotein receptor-related protein 4; LRP10; Lrp4; LRP-4; mdig; Megf7; multiple epidermal growth factor-like domains 7; SOST2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
4038 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O75096 | |
LRP4 | |
Synthetic peptide sequence (SNPSYRTSTQEVK ) corresponding to the internal amino acids of LRP4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |