missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lysozyme Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA129680
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Predigestion with a proteolytic enzyme improves staining of formalin fixed paraffin embedded tissue sections.
Lysozyme Polyclonal antibody specifically detects Lysozyme in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Lysozyme | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P61626 | |
LYZ | |
Purified human lysozyme from urine of patients with monocystic leukemia. | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
LYZ | |
1,4-beta-N-acetylmuramidase C; 1700038F02Rik; 4-beta-N-acetylmuramidase C; AI326280; Allergen Gal d IV; bA534G20.1; c-type lysozyme; dystrophin; egg white lysozyme; Gal d 4; KAAG648; LYC2; Lys; Lysm; lysozyme; lysozyme (renal amyloidosis); lysozyme 1; lysozyme 2; lysozyme C; Lysozyme C type M; lysozyme C type P; Lysozyme C, spleen isozyme; lysozyme C-1; Lysozyme C-2; lysozyme C-3; lysozyme d1; lysozyme F1; lysozyme like 1; lysozyme-like 1; lysozyme-like protein 1; lysozyme-like protein 2; Lysz; LYZ; Lyz1; Lyz2; LYZC; LYZD1; LYZF1; LYZL1; Lyzs; Lzm; Lzm-s1; Lzp; Lzp-s; P lysozyme structural; PRO1278; renal amyloidosis; RGD1559869; unnamed protein product; UNQ648/PRO1278 | |
Rabbit | |
IgG fraction | |
RUO | |
4069 | |
4° C, do not freeze | |
Liquid |