missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOX Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA146020
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Lysyl oxidase (LOX), a copper-containing amine oxidase, belongs to a heterogeneous family of enzymes that oxidize primary amine substrates to reactive aldehydes. It plays a vital role in the formation and repair of the extra cellular matrix. As well, LOX is a multifunctional enzyme having diverse biological functions such as developmental regulation, tumor suppression, cell motility and cellular senescence. The secreted form of LOX is responsible for the invasive properties of hypoxic human cancer cells. Thus, it is essential for hypoxia-induced metastasis and is a good therapeutic target for preventing and treating metastases.Spezifikation
LOX | |
Polyclonal | |
LOX | |
LOX , lysyl oxidase , MGC105112 , protein-lysine 6-oxidase | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
Unconjugated | |
P28300, P28301, P16636 | |
LOX | |
Two synthetic peptides (residues 150-200 and 275-325) of the rat LOX protein. | |
RUO | |
16948, 24914, 4015 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |