Learn More
KARS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527620
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: H1299. Predicted reactivity: Mouse (94%), Rat (93%), Zebrafish (85%), Xenopus laevis (88%), Chicken (86%), Rhesus Monkey (96%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. Lysyl-tRNA synthetase is a homodimer localized to the cytoplasm which belongs to the class II family of tRNA synthetases. It has been shown to be a target of autoantibodies in the human autoimmune diseases, polymyositis or dermatomyositis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Spezifikation
KARS | |
Polyclonal | |
Unconjugated | |
KARS1 | |
AA589550; AL024334; AL033315; AL033367; CMTRIB; D8Ertd698e; D8Wsu108e; DFNB89; EC 6.1.1.6; KARS; Kars1; KARS2; KIAA0070; KRS; lysine tRNA ligase; lysine--tRNA ligase; LysRS; lysyl-tRNA synthetase; mKIAA0070 | |
Rabbit | |
Protein A | |
RUO | |
3735, 85305 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 40% glycerol and 0.01% thimerosal; pH 7 | |
Q15046, Q99MN1 | |
KARS1 | |
Recombinant fragment corresponding to a region within amino acids 280 and 597 of Human Lysyl-tRNA synthetase | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |