Learn More
anti-Mel-CAM/CD146 Oligoclonal, Invitrogen™
Mel-CAM/CD146Oligoclonal antibody 710230 from Thermo Scientific™ Pierce™ specifically detects Mel-CAM/CD146 in Human samples, and it is validated for WB
Marke: Invitrogen
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MUC18, also known as CD146 is an integral membrane glycoprotein belonging to the immunoglobulin superfamily. The protein is expressed at cell-cell junctions in all endothelial cells, and has also been observed in other normal and malignant cell types. In addition to its recognized role as a calcium-independent cell adhesion molecule, MUC18 may also act as a signal transduction molecule in the recruitment of Fyn kinase, and in the subsequent tyrosine-phosphorylation of intracellular proteins involved in actin cytoskeleton assembly.
710230 detects Mel-CAM/CD146 in Human samples. 710230 has been successfully used in WB procedures. The immunogen for 710230 is peptide corresponding to amino acids 375-386 of human melanoma cell adhesion molecule.
Applications:
Western Blotting
Spezifikation
Mel-CAM/CD146 | |
2HCLC | |
P43121 | |
Peptide corresponding to amino acids 375-386 of human melanoma cell adhesion molecule | |
Primary | |
Human | |
Purified |
Polyclonal | |
Unconjugated | |
Rabbit | |
Protein A | |
4162 | |
Store at 2 to 8°C for up to 1 month. For long term storage, store at -20°C. | |
IgG |