Learn More
MOBP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA545545
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: SQKPAKEGPR LSKNQKYSEH FSIHCCPPFT FLNSKKEIVD RKYSICKSGC Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 85%; Horse: 92%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 86%; Rat: 93%
MOBP Polyclonal antibody specifically detects MOBP in Human samples. It is validated for Western Blot.Spezifikation
MOBP | |
Polyclonal | |
Unconjugated | |
Mobp | |
AI835053; AI839848; Mobp; MOBP155; MOBP170; MOBP69; MOBP73; Mobp81; Mobp81p; myelin associated oligodendrocyte basic protein; myelin-associated oligodendrocyte basic protein; myelin-associated oligodendrocytic basic protein; Myelin-associated/Oligodendrocytic Basic Protein-81; R74645; RNMOBP81P; Unknown (protein for MGC:128384) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
4336 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q13875 | |
Mobp | |
Synthetic peptide directed towards the N-terminal of human MOBP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |