missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG Fc Cross-Adsorbed Secondary Antibody, AP, Invitrogen™
Goat Polyclonal Secondary Antibody
Marke: Invitrogen A16093
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Mouse IgG Fc Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG Fc Cross-Adsorbed in Mouse samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Spezifikation
Mouse IgG Fc Cross-Adsorbed | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Goat | |
Affinity chromatography | |
RUO | |
Mouse | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1.5 mg/mL | |
PBS with 0.1mM ZnCl2, 1% BSA, 30mM triethanolamine, 5mM MgCl2 and 0.05% sodium azide; pH 7.2 | |
Gamma Immunoglobin Fc region | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |