Learn More
RORγt Mouse anti-Mouse, Alexa Fluor 647, Clone: Q31-378, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 562682
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The Q31-378 monoclonal antibody recognizes mouse RORgamma t (RORγt), an isoform of RORgamma (RORγ). RORγt is a DNA-binding transcription factor that belongs to the ROR/RZR orphan nuclear receptor family. RORγt is expressed exclusively by lymphoid cells including CD4+CD8+ thymocytes, peripheral CD4+ Th17 and CD8+ Tc17 cells, NKT cells and innate lymphoid cells such as lymphoid tissue inducer (LTi) cells. RORγt plays essential roles in thymopoiesis, T cell homeostasis, differentiation of effector T lymphocytes and the development of secondary lymphoid tissues including lymph nodes and Peyer’s patches.
Spezifikation
RORγt | |
Monoclonal | |
0.2mg/mL | |
RORγT; RORgt; RORgamma t; RORgammaT; Rorc2; Rorg; TOR; Thor; Nr1f3 | |
Mouse | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
Q31-378 | |
Alexa Fluor 647 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse RORγt Recombinant Protein | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |