missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MPST Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528779
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, mouse liver. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MPST Polyclonal antibody specifically detects MPST in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
MPST | |
Polyclonal | |
Unconjugated | |
Mpst | |
3-mercaptopyruvate sulfurtransferase; human liver rhodanese; mercaptopyruvate sulfurtransferase; MPST; MST; testicular tissue protein Li 200; tRNA thiouridin modification protein 1; TST2; TUM1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
246221, 4357 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.57 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P25325, Q99J99 | |
Mpst | |
Synthetic peptide corresponding to a region within amino acids 234 and 297 of Human MPST | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |