Learn More
MRPL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531487
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: A549, HeLa, HCT116. Predicted reactivity: Mouse (90%), Rat (89%), Xenopus laevis (80%), Chicken (80%), Chimpanzee (96%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MRPL3 Polyclonal antibody specifically detects MRPL3 in Human samples. It is validated for Western Blot.Spezifikation
MRPL3 | |
Polyclonal | |
Unconjugated | |
MRPL3 | |
2010320L16Rik; 39S ribosomal protein L3, mitochondrial; 5930422H18Rik; AA407889; COXPD9; L3mt; mitochondrial 60S ribosomal protein L3; Mitochondrial large ribosomal subunit protein uL3m; mitochondrial ribosomal protein L3; MRL3; Mrpl3; MRP-L3; RPML3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11222 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.88 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P09001 | |
MRPL3 | |
Recombinant fragment corresponding to a region within amino acids 150 and 348 of Human MRPL3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |