Learn More
MRPL49 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA545058
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: ESVDEYQFVE RLLPATRIPD PPKHEHYPTP SGWQPPRDPP PNLPYFVRRS Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 79%; Rabbit: 83%; Rat: 86%
MRPL49 Polyclonal antibody specifically detects MRPL49 in Human samples. It is validated for Western Blot.Spezifikation
MRPL49 | |
Polyclonal | |
Unconjugated | |
MRPL49 | |
39S ribosomal protein L49, mitochondrial; 4833419D18Rik; Aa2-277; C11orf4; L49mt; MGC10656; Mitochondrial large ribosomal subunit protein mL49; mitochondrial ribosomal protein L49; Mrpl49; MRP-L49; Neighbor of FAU; neighbor of fau 1; next to FAU; NOF; NOF1; OK/SW-cl.67; Protein NOF1; Unknown (protein for MGC:142600) | |
Rabbit | |
Affinity Chromatography | |
RUO | |
740 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q13405 | |
MRPL49 | |
Synthetic peptide directed towards the N-terminal of human MRPL49 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |