missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MT-ND2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA537185
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
This antibody detects endogenous protein at a molecular weight of 39 kDa. Purity is >95% by SDS-PAGE.
MT-ND2 Polyclonal antibody specifically detects MT-ND2 in Human, Mouse, Rat samples. It is validated for Western Blot.Spezifikation
MT-ND2 | |
Polyclonal | |
Unconjugated | |
MT-ND2 | |
mitochondrially encoded NADH dehydrogenase 2; Mtnd2; mt-Nd2; NADH dehydrogenase subunit 2; NADH2; NADH-ubiquinone oxidoreductase chain 2; ND2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17717, 26194, 4536 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P03891, P03893, P11662 | |
MT-ND2, ND2 | |
A synthetic peptide corresponding to residues in human MT-ND2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |