missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MTGR1 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marke: Invitrogen PA112415
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MTGR1 Polyclonal antibody specifically detects MTGR1 in Human samples. It is validated for Western Blot.Spezifikation
MTGR1 | |
Polyclonal | |
PBS with 0.08% sodium azide | |
O43439 | |
CBFA2T2 | |
C-terminal portion of recombinant MTGR1. | |
250 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CBFA2T2 | |
A430091M07; C330013D05Rik; CBFA2/RUNX1 partner transcriptional co-repressor 2; CBFA2/RUNX1 translocation partner 2; CBFA2T2; CBFA2T2 identified gene homolog; Cbfa2t2h; core-binding factor runt domain alpha subunit 2 translocated to 2 homolog; core-binding factor, runt domain, alpha subunit 2, translocated to, 2 (human); core-binding factor, runt domain, alpha subunit 2, translocated to, 2 homolog; core-binding factor, runt domain, alpha subunit 2; translocated to, 2; DKFZp313F2116; EHT; ETO homolog on chromosome 20; ETO homologous on chromosome 20; MTG8-like protein; MTG8-related protein 1; MTGR1; myeloid translocation gene-related protein 1; myeloid translocation-related protein 1; p85; Protein CBFA2T2; RP5-1137F22.1; translocated to, 2; ZMYND3; ZMYND3 CBFA2T2 | |
Sheep | |
Ammonium sulfate precipitation | |
RUO | |
9139 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |