Learn More
MTMR9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA531188
Detalles adicionales : Netto-Gewicht : 0.01000kg
Descripción
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Human (99%), Mouse (96%), Rat (96%), Zebrafish (85%), Xenopus laevis (83%), Chicken (85%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MTMR9 Polyclonal antibody specifically detects MTMR9 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Especificaciones
MTMR9 | |
Polyclonal | |
Unconjugated | |
Mtmr9 | |
9430075G12Rik; AA516943; AF073881; C8orf9; DKFZP434K171; Inactive phosphatidylinositol 3-phosphatase 9; LIP-STYX; MGC126672; mMTMH3; Mtmr3; MTMR8; MTMR9; myotubularin related protein 8; myotubularin related protein 9; myotubularin-related protein 9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
66036 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96QG7 | |
Mtmr9 | |
Recombinant protein fragment corresponding to a region within amino acids 254 and 428 of Human MTMR9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |