missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC2 Monoclonal Antibody (996/1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA125020
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MUC2 Monoclonal antibody specifically detects MUC2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Spezifikation
MUC2 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Q02817 | |
MUC2 | |
MUC-2 tandem repeat peptide | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
996/1 | |
Unconjugated | |
MUC2 | |
2010015E03Rik; colonic mucin; HH-Muc; intestinal mucin-2; MCM; MLP; muc2; MUC-2; mucin 2; mucin 2, intestinal/tracheal; mucin 2, oligomeric mucus/gel-forming; mucin-2; mucin-like protein; secreted gel-forming mucin; SMUC; wnn | |
Mouse | |
Protein G | |
RUO | |
4583 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |