Learn More
MUC2 Monoclonal Antibody (SPM512), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA138215
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is small intestine tissue.
Secreted epithelial mucins are large macromolecules which exhibit extreme polydispersity. Mucin 2 is the major intestinal mucin. O-glycans are attached to MUC2 in a potentially diverse arrangement, which is crucial for their interaction with endogeneous and exogeneous lectins.Spezifikation
MUC2 | |
Monoclonal | |
Unconjugated | |
MUC2 | |
2010015E03Rik; colonic mucin; HH-Muc; intestinal mucin-2; MCM; MLP; muc2; MUC-2; mucin 2; mucin 2, intestinal/tracheal; mucin 2, oligomeric mucus/gel-forming; mucin-2; mucin-like protein; secreted gel-forming mucin; SMUC; wnn | |
Mouse | |
Protein A/G | |
RUO | |
4583 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin) | |
SPM512 | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
Q02817 | |
MUC2 | |
Synthetic peptide corresponding to a site on the MUC-2 glycoprotein | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG1 |