missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MURF2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518142
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with canine, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. This protein associates transiently with microtubules, myosin, and titin during muscle sarcomere assembly. It may act as a transient adaptor and plays a regulatory role in the assembly of sarcomeres. Four alternatively spliced transcript variants encoding distinct isoforms have been described.Spezifikation
MURF2 | |
Polyclonal | |
Unconjugated | |
TRIM55 | |
D830041C10Rik; MURF2; MuRF-2; muscle specific ring finger 2; muscle-specific RING finger protein 2; RING finger protein 29; RNF29; Trim55; tripartite motif containing 55; tripartite motif-containing 55; tripartite motif-containing protein 55 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
84675 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9BYV6 | |
TRIM55 | |
Synthetic peptide sequence (HIFSFSWLNSLNE) corresponding to the C-terminus amino acids of TRIM55 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |