missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OAS2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA537878
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
OAS2 Polyclonal antibody specifically detects OAS2 in Human samples. It is validated for Western Blot.Spezifikation
OAS2 | |
Polyclonal | |
Unconjugated | |
OAS2 | |
(2-5')oligo(A) synthase 2; (2'-5')oligo(A) synthetase 2; 2',5'-oligoadenylate synthetase-like 11; 2,5-oligoadenylate synthetase-like 11; 2'-5' oligoadenylate synthetase 2; 2'-5' oligoadenylate synthetase-like 11; 2-5A synthase 2; 2'-5'-oligoadenylate synthase 2; 2'-5'-oligoadenylate synthetase 2; 2'-5'-oligoadenylate synthetase 2, 69/71kDa; A synthetase 2; MGC78578; Oas2; Oasl11; p69 OAS / p71 OAS; p69OAS / p71OAS | |
Goat | |
Antigen affinity chromatography | |
RUO | |
4939 | |
-20°C | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P29728 | |
OAS2 | |
Peptide with sequence C-RKTVLRGNSDGT, from the internal region near the N-terminus of OAS2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |