Learn More
OCIAD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA543166
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: QGPDPNLEES PKRKNITYEE LRNKNRESYE VSLTQKTDPS VRPMHERVPK Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 92%; Rabbit: 92%; Rat: 100%
OCIAD1 Polyclonal antibody specifically detects OCIAD1 in Human samples. It is validated for Western Blot.Spezifikation
OCIAD1 | |
Polyclonal | |
Unconjugated | |
Ociad1 | |
6030432N09Rik; AI481327; ASRIJ; AW557942; B230209J16Rik; BB021357; Emi2; expressed during mesenchymal induction 2; FLJ20455; Imi2; induced during mesenchymal induction 2; LOC100723738; MGC111072; OCIA; OCIA domain containing 1; OCIA domain-containing protein 1; Ociad1; OCIAD1 protein; Ovarian cancer immunoreactive antigen domain containing 1; Ovarian carcinoma immunoreactive antigen; TPA018 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54940 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9NX40 | |
Ociad1 | |
Synthetic peptide directed towards the C-terminal of human OCIAD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |