missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCIAD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA520835
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
A suggested positive control is A549 cell lysate. PA5-20835 can be used with blocking peptide PEP-0949.
OCIAD2 Polyclonal antibody specifically detects OCIAD2 in Human samples. It is validated for Western Blot.Spezifikation
OCIAD2 | |
Polyclonal | |
Unconjugated | |
OCIAD2 | |
1810027I20Rik; OCIA domain containing 2; OCIA domain-containing protein 2; OCIAD2; Ovarian carcinoma immunoreactive antigen-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
132299 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q56VL3 | |
OCIAD2 | |
An 18 amino acid peptide from near the carboxy terminus of human OCIAD2. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |