missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OMP Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen OSG00002G
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The peptide is homologous with the corresponding sequence derived from OMP protein in mouse.
Olfactory marker protein (OMP) is an abundant, 19-kDa, cytosolic protein that is almost exclusively expressed in mature, functioning, olfactory neurons but not in the neural precursor basal cells. The OMP gene structure and protein sequence are highly conserved between mouse, rat and human. Its tissue-specific expression in the receptor cells together with the results of the mouse knockout studies show that OMP represents a novel modulatory component of the odor detection/signal transduction cascade. May act as a modulator of the olfactory signal-transduction cascade. Interacts with BEX1 and BEX2. Uniquely associated with mature olfactory receptor neurons. Belongs to the olfactory marker protein family.Spezifikation
OMP | |
Polyclonal | |
Omp | |
Olfactory neuronal-specific protein | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
18378, 24612 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Q64288, P08523 | |
OMP | |
Synthetic peptide from rat Olfactory Marker Protein. | |
500 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |