missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LS189744 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA132773
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-32773 detects Olfactory receptor LS189744 from human samples.
LS189744 Polyclonal antibody specifically detects LS189744 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
LS189744 | |
Polyclonal | |
Unconjugated | |
OR6K3 | |
OR6K3 | |
Synthetic 18 amino acid peptide from C-terminus of human OR6K3 | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q8NGY3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
391114 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |