missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olfr156 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute with 100 μL of distilled water. Centrifuge to remove any insoluble material. OSR00044W can be used with blocking peptide GSM00012P.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.Spezifikation
Olfr156 | |
Polyclonal | |
Olfr156 | |
OLFR156 | |
Synthetic peptide from mouse olfactory receptor family 262, subfamily 6, 3, 5, 7, 11 and 12. | |
RUO | |
29846 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Olfactory receptor 156, MOR262-3, MOR262-5, MOR262-7, MOR262-11, MOR262-12, Olfactory receptor MOR262-6, Olfactory receptor 155, Olfr156, Olfr157, Olf, MOR262-6, MOR262- 3, MOR262- 5, MOR262- 7, MOR262- 11, MOR262- 12 | |
Rabbit | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |