missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olfr620/Olfr624 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSR00198W
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
OSR00198W is specific for MOR4-1 (OLFR620) and MOR4-2P (OLFR624).
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome.Spezifikation
Olfr620/Olfr624 | |
Polyclonal | |
Olfr620 | |
OLFR620, OLFR624 | |
Synthetic peptide from mouse olfactory receptor family 4, subfamily 1 and 2P. | |
RUO | |
258189, 258808 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Olfr620, Olfr624, Olfactory receptors Olfr620, MOR4-1, Olfr624, MOR4-2P | |
Rabbit | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |