Learn More
OMD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530977
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Mouse (83%), Rat (82%), Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
OMD Polyclonal antibody specifically detects OMD in Human samples. It is validated for Western Blot.Spezifikation
OMD | |
Polyclonal | |
Unconjugated | |
OMD | |
Keratan sulfate proteoglycan osteomodulin; KSPG osteomodulin; Omd; OSAD; Osteoadherin; osteoadherin proteoglycan; osteomodulin; SLRR2C; UNQ190/PRO216; UNQ190/PRO216SLRR2C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4958 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q99983 | |
OMD | |
Recombinant fragment corresponding to a region within amino acids 1 and 260 of Human OMD | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |