missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neisseria meningitidis Orf1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA119444
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Western Blot: Reducing conditions.
Neisseria meningitidis Orf1 Polyclonal antibody specifically detects Neisseria meningitidis Orf1 in Bacteria samples. It is validated for Western Blot.Spezifikation
Neisseria meningitidis Orf1 | |
Polyclonal | |
Unconjugated | |
Meningococcus | |
Slices containing purified FrpD250-6xHis-tagged protein were excised from 10% SDS-PAGE gels, grinded, mixed with 1 mg/ml of aluminum hydroxide in PBS. | |
100 μg | |
Primary | |
4° C, do not freeze | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Bacteria | |
Antibody | |
IgG |