Learn More
Osteocalcin Monoclonal Antibody (6F9G4E10), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA182973
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody does not react with bovine, mouse or rat. Mouse anti Human Osteocalcin antibody, clone 6F9G4E10 recognizes intact human osteocalcin in a 2 site assay and reacts with osteocalcin fragments corresponding to amino acids 1-19, 7-19 and 15-31 of the native molecule. It does not recognize fragments representing amino acids 20-42 or intact bovine osteocalcin.
Osteocalcin Monoclonal antibody specifically detects Osteocalcin in Human samples. It is validated for ELISA.Spezifikation
Osteocalcin | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P02818 | |
BGLAP | |
Recombinant human osteocalcin GST fusion protein | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
6F9G4E10 | |
Unconjugated | |
BGLAP | |
Bglap; Bglap1; Bglap2; BGP; Bgpr; Bgpra; bone gamma carboxyglutamate protein; bone gamma carboxyglutamate protein 1; bone gamma-carboxyglutamate (gla) protein; bone gamma-carboxyglutamate (gla) protein (osteocalcin); bone gamma-carboxyglutamate protein; bone gamma-carboxyglutamate protein 2; bone gamma-carboxyglutamate protein osteocalcin; bone gamma-carboxyglutamate; BGLAP; bone gamma-carboxyglutamic acid (Gla) protein (osteocalcin); bone Gla protein; bone gla protein BGP; bone Gla-protein; gamma-carbooxyglutamate (gla) protein; Gamma-carboxyglutamic acid-containing protein; GLA; Gla protein precusor; mOC-A; OC; OCN; OG1; Osteocalcin; PMF1; RP11-54H19.5; unnamed protein product | |
Mouse | |
Protein A | |
RUO | |
632 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |