Learn More
OVOL1 Monoclonal Antibody (1B3), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA519515
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide Sequence: MPRAFLVKKP CVSTCKRNWS ELPDEGRGEI YVPVSLGFCP PQPYREPEPS VAEPPSCPLA LNMSLRDSSY SMAPGPCVVA QLPSEDMGHL TDPQSRDHGF LRAKMKVTLG DSPSGDLFTC RVCQRAFTYQ RMLNRHMKCH NDVKRHLCTY CGKGFNDTFD LKRHVRTHTG VRPYKCSLCD KAFTQRCSLE SHLKKIHGVQ QKYAYKERRA KLYVCEECGC TSESQEGHVL HLKEHHPDSP LLRKTSKKVA VALQNTVTSL LQGSPHL
OVOL1 Monoclonal antibody specifically detects OVOL1 in Human samples. It is validated for ELISA.Spezifikation
OVOL1 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
O14753 | |
OVOL1 | |
OVOL1 (AAH59408, 1 a.a. ∽ 267 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA | |
1B3 | |
Unconjugated | |
OVOL1 | |
BB147136; CD254; hOvo1; hRANKL2; Ly109l; movo1; mOvo1a; ODF; OPG; OPG ligand; OPGL; OPTB2; osteoclast differentiation factor; Osteoprotegerin ligand; OVO homolog-like 1; OVO homolog-like 1 (Drosophila); ovo like transcriptional repressor 1; ovo like zinc finger 1; Ovo1; Ovol1; OVO-like 1 binding protein; ovo-like 1(Drosophila); ovo-like zinc finger 1; Putative transcription factor Ovo-like 1; RANKL; receptor activator of NF-kappaB ligand; receptor activator of nuclear factor kappa B ligand; receptor activator of nuclear factor kappa-B ligand; sOdf; TNF-related activation-induced cytokine; TNFSF11; TNLG6B; Trance; transcription factor Ovo-like 2; tumor necrosis factor (ligand) superfamily member 11; tumor necrosis factor (ligand) superfamily, member 11; tumor necrosis factor ligand 6B; tumor necrosis factor ligand superfamily member 11; Tumor necrosis factor ligand superfamily member 11, membrane form; Tumor necrosis factor ligand superfamily member 11, soluble form; tumor necrosis fact | |
Mouse | |
Affinity chromatography | |
RUO | |
5017 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |