Learn More
P-Cadherin Mouse, Unlabeled, Clone: 56, BD
Mouse Monoclonal Antibody
330.00 € - 545.00 €
Spezifikation
Antigen | P-Cadherin |
---|---|
Klon | 56 |
Konzentration | 250μg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15805349
|
BD Biosciences
610227 |
50 μg |
330.00 €
1 Set |
Verfügbar ab: 05-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15815349
|
BD Biosciences
610228 |
150 μg |
545.00 €
1 Set |
Verfügbar ab: 29-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Cadherins are a family of transmembrane glycoproteins involved in the Ca2+-dependent cell-cell adhesion that occurs in many tissues. Cadherins are similar in their domain structure, Ca2+ and protease sensitivity, and molecular weight. However, these proteins have distinct tissue expression patterns and immunological reactivities. P-Cadherin is localized in placenta while E-Cadherin and N-Cadherin are found in epithelial and neural tissues, respectively. P-Cadherin is an 829 amino acid polypeptide with a putative signal peptide and precursor region, an extracellular domain containing several internal repeats, and a highly hydrophobic transmembrane region. The cytoplasmic domain provides a link to the cytoskeleton through the associated catenin proteins.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Spezifikation
P-Cadherin | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
56 | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.