Learn More
p14ARF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA532529
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is lymphoma.
p14ARF Polyclonal antibody specifically detects p14ARF in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Spezifikation
p14ARF | |
Polyclonal | |
PBS with 1% BSA and 0.1% sodium azide; pH 7.6 | |
P42771 | |
CDKN2A | |
Synthetic peptide derived from C-terminus of human p14 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
CDKN2A | |
Alternative reading frame; ARF; ARF-INK4a; CDK4 inhibitor p16-INK4; CDK4I; CDKN2; CDKN2A; cdkn2a {ECO:0000312; cell cycle inhibitor; cell cycle negative regulator beta; cell cycle regulator; CMM2; cyclin dependent kinase inhibitor 2A; Cyclin dependent kinase inhibitor 2A (p16, inhibits CDK4); cyclin-dependent kinase 4 inhibitor A; Cyclin-dependent kinase inhibitor 2A; cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2A (p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2a p16Ink4a; cyclin-dependent kinase inhibitor 2a p19Arf; cyclin-dependent kinase inhibitor 2A, isoform 1; cyclin-dependent kinase inhibitor 2A, isoform 2; cyclin-dependent kinase inhibitor 2A, isoform 3; cyclin-dependent kinase inhibitor 2A, isoforms 1/2; cyclin-dependent kinase inhibitor protein; EMBL:AAL76336.1, ECO:0000312; EMBL:AAL76338.1, ECO:0000312; inhibits CDK4; INK4; INK4A; Ink4a/Arf; INK4a-ARF; mitochondrial smARF; MLM; MTS1; MTS-1; Multiple tumor sup | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1029 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |