missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p300 Monoclonal Antibody (RW128), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA116608
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
Suggested positive control: HeLa nuclear extract.
p300 Monoclonal antibody specifically detects p300 in Human, Mouse, Mustelid, Non-human Primate, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot.Spezifikation
p300 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
B2RWS6, Q09472 | |
EP300 | |
Fusion protein containing residues 1572-2371 of human p300. | |
100 μL | |
Primary | |
Human, Mouse, Mustelid, Non-human Primate, Rat | |
Antibody | |
IgG1 κ |
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
RW128 | |
Unconjugated | |
EP300 | |
A430090G16; A730011L11; E1A binding protein p300; E1A-associated protein p300; E1A-binding protein, 300kD; Ep300; Histone acetyltransferase p300; Histone butyryltransferase p300; Histone crotonyltransferase p300; KAT3B; P300; p300 HAT; Protein propionyltransferase p300; RSTS2 | |
Mouse | |
Protein A | |
RUO | |
170915, 2033, 328572 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |